ENA321765078 Enamine

2-(3-nitrophenyl)-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD12818488
  • Cas Number : 92057-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76087
Availability details for
Quantity
  • Availability
  • Add to Cart