ENA321765081 Enamine

{2-[4-(benzyloxy)phenoxy]ethyl}(methyl)amine

  • MDL number MFCD12463546
  • Cas Number : 1001587-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77337
Availability details for
Quantity
  • Availability
  • Add to Cart