ENA321765082 Enamine

6-chloro-1-phenyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD18917258
  • Cas Number : 6288-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280-281
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76489
Availability details for
Quantity
  • Availability
  • Add to Cart