ENA321765083 Enamine

5-(1-phenyl-1H-pyrazol-4-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11168104
  • Cas Number : 1152906-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283...285
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76089
Availability details for
Quantity
  • Availability
  • Add to Cart