ENA321765087 Enamine

4-(2-methyl-1H-imidazol-1-yl)pyridin-3-amine

  • MDL number MFCD11184550
  • Cas Number : 1040314-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77345

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77345
Availability details for
Quantity
  • Availability
  • Add to Cart