ENA321765090 Enamine

methyl 1-oxo-1,2-dihydroisoquinoline-3-carboxylate

  • MDL number MFCD10655727
  • Cas Number : 69454-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77350

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77350
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart