ENA321765096 Enamine

3,5-diethyl-1H-1,2,4-triazole

  • MDL number MFCD14620080
  • Cas Number : 7343-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77357
Availability details for
Quantity
  • Availability
  • Add to Cart