ENA321765099 Enamine

9-chloro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD03695449
  • Cas Number : 141106-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77360
Availability details for
Quantity
  • Availability
  • Add to Cart