ENA321765100 Enamine

N-methyl-1,2,3,4-tetrahydronaphthalen-2-amine

  • MDL number MFCD09835327
  • Cas Number : 19485-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77361

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77361
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart