ENA321765115 Enamine

{2-methoxy-5-[(methylsulfanyl)methyl]phenyl}boranediol

  • MDL number MFCD14687399
  • Cas Number : 1334684-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77376

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77376
Availability details for
Quantity
  • Availability
  • Add to Cart