ENA321765119 Enamine

N-[4-(2,5-dioxoimidazolidin-4-yl)phenyl]acetamide

  • MDL number MFCD09739473
  • Cas Number : 22706-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76507
Availability details for
Quantity
  • Availability
  • Add to Cart