ENA321765127 Enamine

4-(2-oxopropyl)morpholine-3,5-dione

  • MDL number MFCD16164344
  • Cas Number : 1248462-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-76509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76509
Availability details for
Quantity
  • Availability
  • Add to Cart