ENA321765134 Enamine

2-bromo-5-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD07783823
  • Cas Number : 141113-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77416
Availability details for
Quantity
  • Availability
  • Add to Cart