ENA321765140 Enamine

4-(quinolin-4-yloxy)aniline

  • MDL number MFCD13438140
  • Cas Number : 863503-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76520
Availability details for
Quantity
  • Availability
  • Add to Cart