ENA321765148 Enamine

2-[4-bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD04969233
  • Cas Number : 1006319-26-1
  • Molecular Weight : 273.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-77441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77441
Availability details for
Quantity
  • Availability
  • Add to Cart