ENA321765154 Enamine

but-3-yn-1-amine hydrochloride

  • MDL number MFCD06658391
  • Cas Number : 88211-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76524

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-76524
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart