ENA321765155 Enamine

1-(3-methyl-1H-pyrazol-5-yl)methanamine

  • MDL number MFCD08059768
  • Cas Number : 479500-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76525
Availability details for
Quantity
  • Availability
  • Add to Cart