ENA321765159 Enamine

6-(3-methylphenyl)-2-sulfanyl-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD02854523
  • Cas Number : 1271475-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-76526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76526
Availability details for
Quantity
  • Availability
  • Add to Cart