ENA321765165 Enamine

[2-(cyclopentyloxy)-3-methoxyphenyl]methanamine

  • MDL number MFCD12177214
  • Cas Number : 1145680-28-9
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76530
Availability details for
Quantity
  • Availability
  • Add to Cart