ENA321765175 Enamine

3-(dimethylcarbamoyl)-4-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD12827497
  • Cas Number : 1114594-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77474

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77474
Availability details for
Quantity
  • Availability
  • Add to Cart