ENA321765176 Enamine

1-benzyl-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD09674969
  • Cas Number : 18159-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77475
Availability details for
Quantity
  • Availability
  • Add to Cart