ENA321765177 Enamine

ethyl 5-amino-1-(2-methylphenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD07388490
  • Cas Number : 1355844-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76532
Availability details for
Quantity
  • Availability
  • Add to Cart