ENA321765201 Enamine

2-(cyclopentylmethyl)butanoic acid

  • MDL number MFCD14694965
  • Cas Number : 14276-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77516
Availability details for
Quantity
  • Availability
  • Add to Cart