ENA321765205 Enamine

2-(cyclohexylmethyl)butanoic acid

  • MDL number MFCD12794552
  • Cas Number : 110604-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77524
Availability details for
Quantity
  • Availability
  • Add to Cart