ENA321765206 Enamine

3-cyclohexyl-2-methylpropanoic acid

  • MDL number MFCD12794494
  • Cas Number : 51953-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77525
Availability details for
Quantity
  • Availability
  • Add to Cart