ENA321765213 Enamine

N-(4-iodophenyl)-3-oxobutanamide

  • MDL number MFCD00053248
  • Cas Number : 38418-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77536
Availability details for
Quantity
  • Availability
  • Add to Cart