ENA321765227 Enamine

1-(2,2,3,3-tetrafluoropropyl)piperazine

  • MDL number MFCD08696506
  • Cas Number : 23476-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77557
Availability details for
Quantity
  • Availability
  • Add to Cart