ENA321765229 Enamine

2,5-dimethoxy-N-methylaniline

  • MDL number MFCD11152066
  • Cas Number : 10224-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77559
Availability details for
Quantity
  • Availability
  • Add to Cart