ENA321765239 Enamine

N-[2-(diethylamino)ethyl]-4-hydroxybenzamide

  • MDL number MFCD00088476
  • Cas Number : 106018-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76571

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76571
Availability details for
Quantity
  • Availability
  • Add to Cart