ENA321765240 Enamine

1-methanesulfonylpropan-2-amine hydrochloride

  • MDL number MFCD15176476
  • Cas Number : 1158368-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76574
Availability details for
Quantity
  • Availability
  • Add to Cart