ENA321765241 Enamine

2-methyl-5-(piperidine-1-sulfonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08445071
  • Cas Number : 100141-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76575
Availability details for
Quantity
  • Availability
  • Add to Cart