ENA321765244 Enamine

1-(hydroxymethyl)pyrrolidine-2,5-dione

  • MDL number MFCD01328751
  • Cas Number : 5146-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76577
Availability details for
Quantity
  • Availability
  • Add to Cart