ENA321765247 Enamine

1-(ethanesulfonyl)propan-2-amine hydrochloride

  • MDL number MFCD15176479
  • Cas Number : 1158485-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76580
Availability details for
Quantity
  • Availability
  • Add to Cart