ENA321765250 Enamine

1-(butan-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805219
  • Cas Number : 1006451-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77602

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77602
Availability details for
Quantity
  • Availability
  • Add to Cart