ENA321765253 Enamine

1-(1-methyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD08700894
  • Cas Number : 40534-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77611
Availability details for
Quantity
  • Availability
  • Add to Cart