ENA321765254 Enamine

3-chloro-4-hydroxybenzohydrazide

  • MDL number MFCD09809436
  • Cas Number : 39634-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280...282
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77612
Availability details for
Quantity
  • Availability
  • Add to Cart