ENA321765256 Enamine

methyl 2-(carbamoylamino)-4-methylpentanoate

  • MDL number MFCD12144047
  • Cas Number : 1485760-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77614
Availability details for
Quantity
  • Availability
  • Add to Cart