ENA321765257 Enamine

4-[4-(aminomethyl)-3-methylphenyl]piperazin-2-one

  • MDL number MFCD16717862
  • Cas Number : 1250744-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77615
Availability details for
Quantity
  • Availability
  • Add to Cart