ENA321765259 Enamine

4-[(2-oxopyrrolidin-1-yl)methyl]benzonitrile

  • MDL number MFCD09947664
  • Cas Number : 1017045-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76108

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76108
Availability details for
Quantity
  • Availability
  • Add to Cart