ENA321765261 Enamine

3-acetyl-4-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD10688980
  • Cas Number : 1152590-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77621
Availability details for
Quantity
  • Availability
  • Add to Cart