ENA321765265 Enamine

4,7-dibromo-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD14673145
  • Cas Number : 20780-89-6
  • Molecular Weight : 304.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-77631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77631
Availability details for
Quantity
  • Availability
  • Add to Cart