ENA321765268 Enamine

6-(5-methylfuran-2-yl)-2-sulfanyl-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD13813389
  • Cas Number : 1014623-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-76597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76597
Availability details for
Quantity
  • Availability
  • Add to Cart