ENA321765275 Enamine

2,2-dimethyloctan-3-amine

  • MDL number MFCD11655425
  • Cas Number : 1099683-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77650
Availability details for
Quantity
  • Availability
  • Add to Cart