ENA321765279 Enamine

2-cyclohexyl-1-(pyrimidin-2-yl)ethan-1-one

  • MDL number MFCD16819898
  • Cas Number : 1249454-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77655

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77655
Availability details for
Quantity
  • Availability
  • Add to Cart