ENA321765281 Enamine

[2-(pyrrolidin-1-yl)pyrimidin-4-yl]methanamine

  • MDL number MFCD10697047
  • Cas Number : 944902-04-9
  • Molecular Weight : 178.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76613

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-76613
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart