ENA321765289 Enamine

3-[1-(4-cyanophenyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD13744133
  • Cas Number : 1173293-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76617

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76617
Availability details for
Quantity
  • Availability
  • Add to Cart