ENA321765290 Enamine

2-(cyclohexyloxy)propanoic acid

  • MDL number MFCD12145311
  • Cas Number : 56674-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76618
Availability details for
Quantity
  • Availability
  • Add to Cart