ENA321765308 Enamine

4-(1,3-dioxolan-2-yl)-4-ethylhexan-1-amine

  • MDL number MFCD18838816
  • Cas Number : 105599-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76662
Availability details for
Quantity
  • Availability
  • Add to Cart