ENA321765309 Enamine

N-(2-methoxyethyl)pyridin-4-amine

  • MDL number MFCD11120010
  • Cas Number : 1039993-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76663

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76663
Availability details for
Quantity
  • Availability
  • Add to Cart