ENA321765314 Enamine

1-(2-bromoethoxy)-3-tert-butylbenzene

  • MDL number MFCD11647764
  • Cas Number : 1099115-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76666

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76666
Availability details for
Quantity
  • Availability
  • Add to Cart